siRNA Details
| Gene Symbol: Lgals3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145953.1 | NP_001139425.1 | 5 | 747 | ||
| NM_010705.3 | NP_034835.1 | 5 | 720 | |||
| GenBank mRNA | AK008593.1 | - | 4 | 389 | ||
| AK089249.1 | - | 5 | 664 | |||
| AK149783.1 | - | 5 | 694 | |||
| AK159536.1 | - | 5 | 688 | |||
| J03723.1 | - | 3 | 620 | |||
| X16834.1 | - | 5 | 666 | |||

Lgals3
Genomic Map