siRNA Details
| Gene Symbol: Eif2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136070.1 | NP_001129542.1 | 12 | 1443 | ||
| NM_010709.3 | NP_034839.2 | 12 | 1443 | |||
| GenBank mRNA | AK030312.1 | - | 12 | 1454 | ||
| AK034115.1 | - | 12 | 1415 | |||
| AK036154.1 | - | 12 | 1439 | |||
| AK046539.1 | - | 12 | 1455 | |||
| AK047005.1 | - | 11 | 1288 | |||
| AK082748.1 | - | 12 | 1458 | |||
| AK083638.1 | - | 12 | 1403 | |||
| AK139052.1 | - | 12 | 1443 | |||
| AK161269.1 | - | 12 | 1426 | |||
| AK166085.1 | - | 12 | 1456 | |||
| AK167980.1 | - | 12 | 1439 | |||
| AK168120.1 | - | 12 | 1409 | |||
| AK170252.1 | - | 12 | 1470 | |||
| AK170448.1 | - | 12 | 1455 | |||
| AK171115.1 | - | 12 | 1421 | |||
| BC023709.1 | - | 12 | 1396 | |||
| BC024501.1 | - | 7 | 757 | |||
| BC025036.1 | - | 12 | 1399 | 4935223 | ||
| U58337.1 | - | 12 | 1328 | |||

Eif2d
Genomic Map