siRNA Details
| Gene Symbol: Limk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141813.1 | - | 3 | 357 | ||
| AK164484.1 | - | 3 | 358 | |||
| AK170166.1 | - | 2 | 132 | |||
| AK172083.1 | - | 2 | 116 | |||
| BC063768.1 | - | 3 | 339 | |||
| BY230595.1 | - | 2 | 159 | |||
| CV563459.1 | - | 2 | 116 | |||
| U14166.1 | - | 3 | 380 | |||
| U15159.1 | - | 3 | 355 | |||

Limk1
Genomic Map