siRNA Details
| Gene Symbol: Lipa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111100.1 | NP_001104570.1 | 10 | 1195 | ||
| NM_021460.3 | NP_067435.3 | 10 | 1199 | |||
| GenBank mRNA | AK088659.1 | - | 10 | 1151 | ||
| AK149791.1 | - | 10 | 1159 | |||
| AK154804.1 | - | 10 | 1139 | |||
| AK164007.1 | - | 10 | 1195 | |||
| AK169567.1 | - | 10 | 1165 | |||
| AK178019.1 | - | 1 | 63 | |||
| BC058564.1 | - | 10 | 1147 | 6441278 | ||
| Z31689.1 | - | 9 | 1104 | |||

Lipa
Genomic Map