siRNA Details
Gene Symbol: Lipa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111100.1 | NP_001104570.1 | 4 | 372 | ||
NM_021460.3 | NP_067435.3 | 4 | 376 | |||
GenBank mRNA | AK088659.1 | - | 4 | 328 | ||
AK149791.1 | - | 4 | 336 | |||
AK154804.1 | - | 4 | 318 | |||
AK164007.1 | - | 4 | 372 | |||
AK169567.1 | - | 4 | 342 | |||
AK209823.1 | - | 2 | 62 | |||
BC058564.1 | - | 4 | 324 | 6441278 | ||
Z31689.1 | - | 3 | 281 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
