siRNA Details
| Gene Symbol: Loxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013586.4 | NP_038614.2 | 14 | 2343 | ||
| GenBank mRNA | AF053368.1 | - | 14 | 2374 | ||
| AK030548.1 | - | 14 | 2344 | |||
| AK145189.1 | - | 9 | 1517 | |||
| BC011298.1 | - | 14 | 2337 | 3488978 | ||

Loxl3
Genomic Map