siRNA Details
| Gene Symbol: Loxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013586.4 | NP_038614.2 | 5 | 955 | ||
| GenBank mRNA | AF053368.1 | - | 5 | 986 | ||
| AK030548.1 | - | 5 | 956 | |||
| AK145189.1 | - | 5 | 975 | |||
| BC011298.1 | - | 5 | 949 | 3488978 | ||

Loxl3
Genomic Map