siRNA Details
| Gene Symbol: Lrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008512.2 | NP_032538.2 | 40 | 6878 | ||
| GenBank mRNA | AF367720.1 | - | 40 | 6849 | ||
| AK154429.1 | - | 25 | 3812 | |||
| X67469.1 | - | 40 | 6858 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrp1| Gene Symbol: Lrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008512.2 | NP_032538.2 | 40 | 6878 | ||
| GenBank mRNA | AF367720.1 | - | 40 | 6849 | ||
| AK154429.1 | - | 25 | 3812 | |||
| X67469.1 | - | 40 | 6858 | |||