siRNA Details
| Gene Symbol: Lrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008513.3 | NP_032539.2 | 12 | 2666 | ||
| GenBank mRNA | AF064984.1 | - | 12 | 2656 | ||
| AF077847.1 | - | 12 | 2544 | |||
| AK090382.1 | - | 3 | 372 | |||
| AK147068.1 | - | 12 | 2659 | |||
| AK147231.1 | - | 13 | 2777 | |||
| AK147330.1 | - | 13 | 2777 | |||
| AK220186.1 | - | 12 | 2619 | |||
| BC011374.1 | - | 12 | 2648 | 3154246 | ||
| BC094610.1 | - | 12 | 2655 | |||

Lrp5
Genomic Map