siRNA Details
| Gene Symbol: Lrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008513.3 | NP_032539.2 | 14, 15 | 3355 | ||
| GenBank mRNA | AF064984.1 | - | 14, 15 | 3345 | ||
| AF077847.1 | - | 14, 15 | 3233 | |||
| AK090382.1 | - | 5, 6 | 1033 | |||
| AK147068.1 | - | 14, 15 | 3348 | |||
| AK147231.1 | - | 15, 16 | 3466 | |||
| AK147330.1 | - | 15, 16 | 3466 | |||
| AK220186.1 | - | 14, 15 | 3308 | |||
| BC011374.1 | - | 14, 15 | 3337 | 3154246 | ||
| BC094610.1 | - | 14, 15 | 3342 | |||

Lrp5
Genomic Map