siRNA Details
| Gene Symbol: Lrrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008516.4 | NP_032542.1 | 2 | 1474 | ||
| GenBank mRNA | AK139355.1 | - | 2 | 1432 | ||
| AK139965.1 | - | 2 | 1476 | |||
| AK160841.1 | - | 2 | 1353 | |||
| AK164849.1 | - | 2 | 1433 | |||
| AK173186.1 | - | 1 | 719 | |||
| BC031122.1 | - | 1 | 749 | 4972803 | ||
| BC058701.1 | - | 2 | 1407 | 6853964 | ||
| D45913.1 | - | 2 | 1420 | |||

Lrrn1
Genomic Map