siRNA Details
| Gene Symbol: Lrrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271708.1 | NP_001258637.1 | 3 | 1029 | ||
| NM_001271709.1 | NP_001258638.1 | 2 | 995 | |||
| NM_010733.3 | NP_034863.1 | 2 | 1000 | |||
| GenBank mRNA | AK036316.1 | - | 2 | 898 | ||
| BC069041.1 | - | 2 | 694 | 5698788 | ||
| D49802.1 | - | 2 | 911 | |||

Lrrn3
Genomic Map