siRNA Details
| Gene Symbol: Ltbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010736.3 | NP_034866.1 | 2, 3 | 385 | ||
| GenBank mRNA | AI119599.1 | - | 2, 3 | 385 | ||
| AK146127.1 | - | 2, 3 | 368 | |||
| BC138590.1 | - | 2, 3 | 248 | 8861610 | ||
| BC138591.1 | - | 2, 3 | 248 | 8861611 | ||
| L38423.1 | - | 2, 3 | 385 | |||
| U29173.1 | - | 2, 3 | 369 | |||

Ltbr
Genomic Map