siRNA Details
| Gene Symbol: Ltbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010736.3 | NP_034866.1 | 10 | 1237 | ||
| GenBank mRNA | AK078859.1 | - | 6 | 496 | ||
| AK146127.1 | - | 10 | 1220 | |||
| BC138590.1 | - | 10 | 1100 | 8861610 | ||
| BC138591.1 | - | 10 | 1100 | 8861611 | ||
| L38423.1 | - | 10 | 1237 | |||
| U29173.1 | - | 10 | 1221 | |||

Ltbr
Genomic Map