siRNA Details
| Gene Symbol: Lum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008524.2 | NP_032550.2 | 2 | 1184 | ||
| GenBank mRNA | AK014312.1 | - | 2 | 880 | ||
| AK075737.1 | - | 2 | 883 | |||
| AK085240.2 | - | 2 | 876 | |||
| AK164774.1 | - | 2 | 876 | |||
| BC005550.1 | - | 2 | 862 | 3585672 | ||
| S79461.1 | - | 2 | 865 | |||

Lum
Genomic Map