siRNA Details
| Gene Symbol: Slc2a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130451.3 | NP_569718.1 | 4, 5 | 1685 | ||
| GenBank mRNA | AK052156.1 | - | 4, 5 | 1652 | ||
| AY029579.1 | - | 4, 5 | 1608 | |||
| BC137917.1 | - | 4, 5 | 1603 | 8860936 | ||
| BC137920.1 | - | 4, 5 | 1603 | 8860939 | ||

Slc2a10
Genomic Map