siRNA Details
| Gene Symbol: Recql5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130454.2 | NP_569721.1 | 11 | 2206 | ||
| GenBank mRNA | AB059999.1 | - | 11 | 1720 | ||
| AK049269.1 | - | 6 | 606 | |||
| AK050965.1 | - | 9 | 935 | |||
| AK078833.1 | - | 1 | 1104 | |||
| BC023769.1 | - | 10 | 1498 | |||
| BC053086.1 | - | 11 | 2459 | |||
| BC059000.1 | - | 11 | 2179 | |||
| BC060070.1 | - | 11 | 2459 | |||
| BC066176.1 | - | 11 | 1929 | |||

Recql5
Genomic Map