siRNA Details
| Gene Symbol: Recql5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130454.2 | NP_569721.1 | 8 | 1975 | ||
| GenBank mRNA | AB059999.1 | - | 8 | 1489 | ||
| AK049269.1 | - | 3 | 375 | |||
| AK050965.1 | - | 6 | 704 | |||
| AK083513.1 | - | 8 | 1972 | |||
| BC023769.1 | - | 7 | 1267 | |||
| BC053086.1 | - | 8 | 2228 | |||
| BC059000.1 | - | 8 | 1949 | |||
| BC060070.1 | - | 8 | 2228 | |||
| BC066176.1 | - | 8 | 1698 | |||

Recql5
Genomic Map