siRNA Details
| Gene Symbol: Snx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130796.4 | NP_570614.2 | 2 | 1879 | ||
| GenBank mRNA | AF408408.1 | - | 2 | 1867 | ||
| AK052343.1 | - | 2 | 1840 | |||
| AK076119.1 | - | 2 | 707 | |||
| AK077890.1 | - | 2 | 1473 | |||
| BC040407.1 | - | 2 | 582 | |||
| BC063089.1 | - | 2 | 1879 | 6516923 | ||
| BC137801.1 | - | 2 | 1865 | 8860820 | ||
| BC145951.1 | - | 2 | 1865 | 40131254 | ||

Snx18
Genomic Map