siRNA Details
| Gene Symbol: Ubn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026666.3 | NP_080942.1 | 13, 14 | 2571 | ||
| GenBank mRNA | AK137727.1 | - | 1, 2 | 37 | ||
| AK147571.1 | - | 13, 14 | 2572 | |||
| BC060723.1 | - | 11, 12 | 1499 | |||
| BC098372.1 | - | 11, 12 | 1731 | 30363389 | ||

Ubn1
Genomic Map