siRNA Details
| Gene Symbol: Cd93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010740.3 | NP_034870.1 | 1 | 509 | ||
| GenBank mRNA | AF081789.1 | - | 1 | 518 | ||
| AK028485.1 | - | 1 | 538 | |||
| AK079060.1 | - | 1 | 510 | |||
| AK155060.1 | - | 1 | 411 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd93| Gene Symbol: Cd93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010740.3 | NP_034870.1 | 1 | 509 | ||
| GenBank mRNA | AF081789.1 | - | 1 | 518 | ||
| AK028485.1 | - | 1 | 538 | |||
| AK079060.1 | - | 1 | 510 | |||
| AK155060.1 | - | 1 | 411 | |||