siRNA Details
| Gene Symbol: Idh3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130884.4 | NP_570954.1 | 8 | 743 | ||
| GenBank mRNA | AK215024.1 | - | 2 | 132 | ||
| AK216020.1 | - | 3 | 281 | |||
| AY035212.1 | - | 8 | 724 | |||
| BC009022.1 | - | 8 | 732 | 3257667 | ||
| CN456483.1 | - | 3 | 291 | |||

Idh3b
Genomic Map