siRNA Details
| Gene Symbol: Idh3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130884.4 | NP_570954.1 | 4 | 296 | ||
| GenBank mRNA | AK046059.1 | - | 3 | 416 | ||
| AY035212.1 | - | 4 | 277 | |||
| BC009022.1 | - | 4 | 285 | 3257667 | ||
| BY336999.1 | - | 4 | 296 | |||
| DN176034.1 | - | 4 | 262 | |||

Idh3b
Genomic Map