siRNA Details
| Gene Symbol: Oxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130163.1 | NP_001123635.1 | 3 | 230 | ||
| NM_001130165.1 | NP_001123637.1 | 3 | 325 | |||
| NM_001130166.1 | NP_001123638.1 | 5 | 589 | |||
| NM_130885.2 | NP_570955.1 | 3 | 230 | |||
| GenBank mRNA | AF324899.1 | - | 3 | 229 | ||
| AF333985.1 | - | 3 | 230 | |||
| AK040881.1 | - | 3 | 252 | |||
| AK052714.1 | - | 3 | 395 | |||
| AK158703.1 | - | 5 | 571 | |||
| BC089183.1 | - | 3 | 320 | 5321255 | ||
| BC098491.1 | - | 5 | 572 | 30086857 | ||

Oxr1
Genomic Map