siRNA Details
| Gene Symbol: Papln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205343.1 | NP_001192272.1 | 21, 22 | 2999 | ||
| NM_130887.3 | NP_570957.2 | 20, 21 | 2933 | |||
| GenBank mRNA | AF314171.1 | - | 20, 21 | 2906 | ||
| AK053198.1 | - | 21, 22 | 3001 | |||
| AK086767.1 | - | 20, 21 | 2916 | |||
| BC005747.1 | - | 2, 3 | 470 | |||
| BC132475.1 | - | 20, 21 | 2873 | 40130752 | ||
| BC137854.1 | - | 20, 21 | 2873 | 8860873 | ||
| BC144975.1 | - | 21, 22 | 2939 | 9053523 | ||

Papln
Genomic Map