siRNA Details
| Gene Symbol: Zfp287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133208.2 | NP_573471.2 | 5 | 1188 | ||
| GenBank mRNA | AF281141.1 | - | 5 | 1188 | ||
| AK029572.1 | - | 5 | 768 | |||
| BC023790.1 | - | 3 | 207 | |||
| BC117876.1 | - | 5 | 913 | 40095930 | ||
| BC117877.1 | - | 5 | 913 | 40095933 | ||

Zfp287
Genomic Map