siRNA Details
| Gene Symbol: Zfp287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133208.2 | NP_573471.2 | 1 | 646 | ||
| GenBank mRNA | AF281141.1 | - | 1 | 646 | ||
| AK029572.1 | - | 1 | 226 | |||
| AK031950.1 | - | 1 | 2497 | |||
| AK046006.1 | - | 2 | 678 | |||
| AK133603.1 | - | 2 | 659 | |||
| AK134983.1 | - | 1 | 224 | |||
| BC117876.1 | - | 1 | 371 | 40095930 | ||
| BC117877.1 | - | 1 | 371 | 40095933 | ||

Zfp287
Genomic Map