siRNA Details
| Gene Symbol: Xpnpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133216.3 | NP_573479.3 | 4 | 395 | ||
| GenBank mRNA | AF363970.1 | - | 4 | 335 | ||
| AK146033.1 | - | 4 | 375 | |||
| AK149676.1 | - | 4 | 334 | |||
| AK158597.1 | - | 2 | 353 | |||
| AK166715.1 | - | 4 | 295 | |||
| AK169024.1 | - | 3 | 221 | |||
| AK188589.1 | - | 2 | 92 | |||
| BC065174.1 | - | 4 | 283 | 6415314 | ||

Xpnpep1
Genomic Map