siRNA Details
| Gene Symbol: Xpnpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133216.3 | NP_573479.3 | 10 | 1048 | ||
| GenBank mRNA | AF363970.1 | - | 10 | 988 | ||
| AK052094.1 | - | 2 | 190 | |||
| AK146033.1 | - | 10 | 1028 | |||
| AK149676.1 | - | 10 | 987 | |||
| AK158597.1 | - | 8 | 1006 | |||
| AK166715.1 | - | 10 | 948 | |||
| AK169024.1 | - | 9 | 874 | |||
| AK177494.1 | - | 1 | 34 | |||
| AK185825.1 | - | 1 | 35 | |||
| BC065174.1 | - | 10 | 936 | 6415314 | ||

Xpnpep1
Genomic Map