siRNA Details
| Gene Symbol: Epcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008532.2 | NP_032558.2 | 8 | 1130 | ||
| GenBank mRNA | AK133510.1 | - | 7 | 812 | ||
| AK145752.1 | - | 7 | 706 | |||
| AK167182.1 | - | 7 | 775 | |||
| BC005618.1 | - | 7 | 771 | 3711069 | ||
| BC094465.1 | - | 7 | 796 | 4948935 | ||
| M76124.1 | - | 7 | 779 | |||

Epcam
Genomic Map