siRNA Details
| Gene Symbol: Hdac10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199198.2 | NP_954668.2 | 5, 6 | 747 | ||
| GenBank mRNA | AK040235.1 | - | 5, 6 | 699 | ||
| AK045028.1 | - | 4, 5 | 958 | |||
| BC013700.1 | - | 5, 6 | 700 | |||
| BC064018.1 | - | 5, 6 | 674 | 3980699 | ||
| BU706947.1 | - | 5, 6 | 488 | |||

Hdac10
Genomic Map