siRNA Details
| Gene Symbol: Crb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133239.2 | NP_573502.2 | 6 | 2256 | ||
| GenBank mRNA | AF406641.1 | - | 6 | 2221 | ||
| AF492496.1 | - | 5 | 2097 | |||
| AK044345.1 | - | 2 | 1095 | |||
| AK044704.1 | - | 2 | 1134 | |||
| AY450552.1 | - | 6 | 2221 | |||
| BC053331.1 | - | 1 | 499 | |||

Crb1
Genomic Map