siRNA Details
| Gene Symbol: Mlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133241.2 | NP_573504.1 | 5 | 458 | ||
| GenBank mRNA | AF449425.1 | - | 5 | 912 | ||
| AK030396.1 | - | 5 | 457 | |||
| AK043757.1 | - | 5 | 458 | |||
| AK122190.1 | - | 5 | 900 | |||
| BC024719.1 | - | 5 | 458 | 5363238 | ||

Mlc1
Genomic Map