siRNA Details
| Gene Symbol: Ms4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133246.5 | NP_573509.1 | 6 | 718 | ||
| GenBank mRNA | AF321128.1 | - | 6 | 657 | ||
| AK036464.1 | - | 6 | 720 | |||
| AY184359.1 | - | 6 | 672 | |||
| BC117013.1 | - | 6 | 634 | 40126332 | ||
| BC117015.1 | - | 6 | 634 | 40126334 | ||
| BC144853.1 | - | 6 | 634 | 9053401 | ||

Ms4a3
Genomic Map