siRNA Details
| Gene Symbol: Vgll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133251.1 | NP_573514.1 | 4, 5 | 529 | ||
| GenBank mRNA | AB036708.1 | - | 4, 5 | 529 | ||
| AK146118.1 | - | 3, 4 | 400 | |||
| BC144928.1 | - | 4, 5 | 477 | 9053476 | ||
| BC145949.1 | - | 3, 4 | 461 | 40131252 | ||

Vgll1
Genomic Map