siRNA Details
| Gene Symbol: Tram2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177409.3 | NP_803128.1 | 9 | 963 | ||
| GenBank mRNA | AK082751.1 | - | 9 | 964 | ||
| AY029530.1 | - | 9 | 900 | |||
| BC018212.1 | - | 9 | 948 | 3980004 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tram2| Gene Symbol: Tram2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177409.3 | NP_803128.1 | 9 | 963 | ||
| GenBank mRNA | AK082751.1 | - | 9 | 964 | ||
| AY029530.1 | - | 9 | 900 | |||
| BC018212.1 | - | 9 | 948 | 3980004 | ||