siRNA Details
| Gene Symbol: Fut10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012517.5 | NP_001012535.1 | 4 | 1357 | ||
| NM_001286422.1 | NP_001273351.1 | 5 | 1462 | |||
| NM_001286424.1 | NP_001273353.1 | 4 | 1250 | |||
| NM_134161.3 | NP_598922.1 | 4 | 1357 | |||
| GenBank mRNA | AJ880009.1 | - | 3 | 970 | ||
| AK147136.1 | - | 5 | 1418 | |||
| AK169153.1 | - | 4 | 1311 | |||
| BC030474.1 | - | 4 | 1254 | 5368333 | ||
| BC062113.1 | - | 4 | 1356 | 6816067 | ||

Fut10
Genomic Map