siRNA Details
| Gene Symbol: Mad1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010752.3 | NP_034882.2 | 14 | 1525 | ||
| GenBank mRNA | AF083812.1 | - | 14 | 1532 | ||
| AF083813.1 | - | 14 | 1532 | |||
| AK136981.1 | - | 14 | 1528 | |||
| AK169976.1 | - | 3 | 494 | |||
| BC010702.1 | - | 14 | 1523 | 3156639 | ||

Mad1l1
Genomic Map