siRNA Details
| Gene Symbol: Galnt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134189.2 | NP_598950.2 | 7 | 1157 | ||
| GenBank mRNA | AK033515.1 | - | 6 | 815 | ||
| AK131155.1 | - | 7 | 1135 | |||
| AK143714.1 | - | 6 | 988 | |||
| AK159088.1 | - | 7 | 1156 | |||
| AK159362.1 | - | 7 | 1155 | |||
| AK162424.1 | - | 7 | 1157 | |||
| BC016585.1 | - | 2 | 269 | 3595220 | ||
| BC060617.1 | - | 7 | 1140 | 6836791 | ||

Galnt10
Genomic Map