siRNA Details
Gene Symbol: Vmn1r226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134191.1 | NP_598952.1 | 1 | 881 | ||
GenBank mRNA | BC119429.1 | - | 1 | 879 | ||
BC119430.1 | - | 1 | 881 | 40040474 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134191.1 | NP_598952.1 | 1 | 881 | ||
GenBank mRNA | BC119429.1 | - | 1 | 879 | ||
BC119430.1 | - | 1 | 881 | 40040474 |