siRNA Details
| Gene Symbol: Vmn1r233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134202.1 | NP_598963.1 | 1 | 657 | ||
| GenBank mRNA | BC115528.1 | - | 1 | 653 | 40101376 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r233| Gene Symbol: Vmn1r233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134202.1 | NP_598963.1 | 1 | 657 | ||
| GenBank mRNA | BC115528.1 | - | 1 | 653 | 40101376 | |