siRNA Details
| Gene Symbol: Vmn1r83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134209.1 | NP_598970.1 | 1 | 772 | ||
| GenBank mRNA | BC125369.1 | - | 1 | 819 | 40129884 | |
| BC138162.1 | - | 1 | 819 | 8861181 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r83| Gene Symbol: Vmn1r83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134209.1 | NP_598970.1 | 1 | 772 | ||
| GenBank mRNA | BC125369.1 | - | 1 | 819 | 40129884 | |
| BC138162.1 | - | 1 | 819 | 8861181 | ||