siRNA Details
Gene Symbol: Vmn1r199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134213.1 | NP_598974.1 | 1 | 696 | ||
GenBank mRNA | BC107236.2 | - | 1 | 696 | 40052151 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134213.1 | NP_598974.1 | 1 | 696 | ||
GenBank mRNA | BC107236.2 | - | 1 | 696 | 40052151 |