siRNA Details
| Gene Symbol: Vmn1r213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134215.1 | NP_598976.1 | 1 | 1043 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r213| Gene Symbol: Vmn1r213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134215.1 | NP_598976.1 | 1 | 1043 | ||