siRNA Details
| Gene Symbol: Vmn1r215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134219.1 | NP_598980.1 | 1 | 611 | ||
| GenBank mRNA | BC141030.1 | - | 1 | 659 | 9055642 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r215| Gene Symbol: Vmn1r215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134219.1 | NP_598980.1 | 1 | 611 | ||
| GenBank mRNA | BC141030.1 | - | 1 | 659 | 9055642 | |