siRNA Details
Gene Symbol: Vmn1r70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134228.1 | NP_598989.1 | 1 | 448 | ||
GenBank mRNA | AF394955.1 | - | 1 | 448 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134228.1 | NP_598989.1 | 1 | 448 | ||
GenBank mRNA | AF394955.1 | - | 1 | 448 |