siRNA Details
| Gene Symbol: Vmn1r82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134234.1 | NP_598995.1 | 1 | 289 | ||
| GenBank mRNA | BC109325.2 | - | 1 | 289 | 40050555 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r82| Gene Symbol: Vmn1r82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134234.1 | NP_598995.1 | 1 | 289 | ||
| GenBank mRNA | BC109325.2 | - | 1 | 289 | 40050555 | |