siRNA Details
| Gene Symbol: Smad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016769.4 | NP_058049.3 | 2 | 641 | ||
| GenBank mRNA | AB008192.1 | - | 2 | 350 | ||
| AF016189.1 | - | 2 | 617 | |||
| AK041743.1 | - | 2 | 162 | |||
| AK048626.1 | - | 2 | 641 | |||
| AK083158.1 | - | 2 | 249 | |||
| BC022904.1 | - | 2 | 784 | |||
| BC066850.1 | - | 2 | 327 | 30432720 | ||

Smad3
Genomic Map