siRNA Details
Gene Symbol: Timd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC025096.1 | - | 5 | 684 | 4158605 | |
BC028829.1 | - | 5 | 716 | 5102129 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Timd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC025096.1 | - | 5 | 684 | 4158605 | |
BC028829.1 | - | 5 | 716 | 5102129 |