siRNA Details
| Gene Symbol: Timd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161355.1 | NP_001154827.1 | 7 | 1014 | ||
| NM_001161356.1 | NP_001154828.1 | 7 | 787 | |||
| NM_134249.5 | NP_599010.4 | 7 | 896 | |||
| GenBank mRNA | AB009015.1 | - | 7 | 787 | ||
| AF399827.1 | - | 7 | 748 | |||
| AF399828.1 | - | 7 | 748 | |||
| AK145630.1 | - | 7 | 775 | |||
| AK163225.1 | - | 7 | 775 | |||
| AK166877.1 | - | 7 | 768 | |||
| BC025096.1 | - | 7 | 826 | 4158605 | ||
| BC028829.1 | - | 7 | 858 | 5102129 | ||

Timd2
Genomic Map